Nickel(II) trifluoromethanesulphonate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Trifluoromethanesulphonic acid Nickel(II) Salt, Nickel(II) Triflate, Nickel(II) trifluoromethanesulphonate, Nickel triflate

PC3387-10G PC3387-25G PC3387-5G PC3387-1G
APOSPC3387-10GEA 0 EUR
APOSPC3387-10G APOSPC3387-25G APOSPC3387-5G APOSPC3387-1G
Nickel(II) trifluoromethanesulphonate
Nickel(II) trifluoromethanesulphonate
Formel: C₂F₆NiO₆S₂
Schmelzpunkt: 100…106 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00673740
CAS-Nummer: 60871-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR