Kaliumvinyltrifluoroborat 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Potassium trifluoro(vinyl)borate

PC3373-25G PC3373-5G PC3373-100G
APOSPC3373-25GEA 81.2 EUR
APOSPC3373-25G APOSPC3373-5G APOSPC3373-100G
Kaliumvinyltrifluoroborat 97%
Kaliumvinyltrifluoroborat
Formel: C₂H₃BF₃K
Schmelzpunkt: 226…230 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02093335
CAS-Nummer: 13682-77-4
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR