5-Fluoropyridine-2-carbonyl chloride
Lieferant: Apollo Scientific
PC32868-1G
PC32868-250MG
PC32868-500MG
APOSPC32868-1GEA
510
EUR
APOSPC32868-1G
APOSPC32868-250MG
APOSPC32868-500MG
5-Fluoropyridine-2-carbonyl chloride
5-Fluoropyridine-2-carbonyl chloride
Formel:
C₆H₃ClFNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13173597 CAS-Nummer: 717871-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...