Ethyl (2,6-difluorophenyl)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(2,6-difluorophenyl)acetate

PC32680-1G PC32680-250MG
APOSPC32680-1GEA 108 EUR
APOSPC32680-1G APOSPC32680-250MG
Ethyl (2,6-difluorophenyl)acetate
Ethyl (2,6-difluorophenyl)acetate
Formel: C₁₀H₁₀F₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01765417
CAS-Nummer: 680217-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR