Ethyl-4-hydroxy-6-(trifluormethoxy)chinolin-3-carboxylat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
PC3247-1G
PC3247-250MG
PC3247-5G
APOSPC3247-1GEA
58.2
EUR
APOSPC3247-1G
APOSPC3247-250MG
APOSPC3247-5G
Ethyl-4-hydroxy-6-(trifluormethoxy)chinolin-3-carboxylat 97%
Ethyl-4-hydroxy-6-(trifluormethoxy)chinolin-3-carboxylat
Formel:
C₁₃H₁₀F₃NO₄ Schmelzpunkt: 297…305 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00219847 CAS-Nummer: 175203-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...