(3-Fluorbenzyl)triphenylphosphoniumchlorid

Lieferant: Apollo Scientific
Achtung

PC31781-1G PC31781-500MG PC31781-5G
APOSPC31781-1GEA 77 EUR
APOSPC31781-1G APOSPC31781-500MG APOSPC31781-5G
(3-Fluorbenzyl)triphenylphosphoniumchlorid
(3-Fluorbenzyl)triphenylphosphoniumchlorid
Formel: C₂₅H₂₁ClFP
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00179768
CAS-Nummer: 55128-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR