2,6-Difluoro-3-nitrobenzamide

Lieferant: Apollo Scientific

PC31534-1G PC31534-250MG PC31534-500MG
APOSPC31534-1GEA 510 EUR
APOSPC31534-1G APOSPC31534-250MG APOSPC31534-500MG
2,6-Difluoro-3-nitrobenzamide
2,6-Difluoro-3-nitrobenzamide
Formel: C₇H₄F₂N₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00120281
CAS-Nummer: 124169-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR