2-Quinolin-2-yl-1-(3-trifluoromethylphenyl)ethanol 95+%
Lieferant: Apollo Scientific
PC3146-25G
PC3146-5G
PC3146-1G
APOSPC3146-25GEA
1260
EUR
APOSPC3146-25G
APOSPC3146-5G
APOSPC3146-1G
2-Quinolin-2-yl-1-(3-trifluoromethylphenyl)ethanol 95+%
2-Quinolin-2-yl-1-(3-trifluoromethylphenyl)ethanol
Formel:
C₁₈H₁₄F₃NO Schmelzpunkt: 103…105 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03424492 CAS-Nummer: 502625-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...