4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol 98%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol

PC302995-10G PC302995-5G
APOSPC302995-10GEA 678 EUR
APOSPC302995-10G APOSPC302995-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol 98%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol
Formel: C₉H₈F₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22201071
CAS-Nummer: 1373921-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR