4-Chlor-3-ethoxy-2-fluorbenzaldehyd 98%
Lieferant: Apollo Scientific
PC302984-10G
PC302984-5G
PC302984-1G
APOSPC302984-10GEA
328
EUR
APOSPC302984-10G
APOSPC302984-5G
APOSPC302984-1G
4-Chlor-3-ethoxy-2-fluorbenzaldehyd 98%
4-Chloro-3-ethoxy-2-fluorobenzaldehyde
Formel:
C₉H₈ClFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD19687180 CAS-Nummer: 1323966-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...