2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl alcohol

PC302919-10G PC302919-5G
APOSPC302919-10GEA 316 EUR
APOSPC302919-10G APOSPC302919-5G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl alcohol 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl alcohol
Formel: C₉H₈F₄O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22201037
CAS-Nummer: 1373921-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR