3'-Fluor-5'-(pentafluorthio)acetophenon 98%
Lieferant: Apollo Scientific
PC302844-1G
PC302844-5G
APOSPC302844-1GEA
348
EUR
APOSPC302844-1G
APOSPC302844-5G
3'-Fluor-5'-(pentafluorthio)acetophenon 98%
3'-Fluor-5'-(pentafluorthio)acetophenon
Formel:
C₈H₆F₆OS Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16652464 CAS-Nummer: 1240257-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...