4-Methoxy-2-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Methoxy-2-(trifluoromethyl)phenyl)methanol

PC302755-10G PC302755-5G PC302755-1G
APOSPC302755-10GEA 249 EUR
APOSPC302755-10G APOSPC302755-5G APOSPC302755-1G
4-Methoxy-2-(trifluormethyl)benzylalkohol 98%
4-Methoxy-2-(trifluormethyl)benzylalkohol
Formel: C₉H₉F₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06202983
CAS-Nummer: 773871-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR