4-Fluoro-3,5-dimethylbenzoic acid 98%
Lieferant: Apollo Scientific
PC302754-5G
PC302754-1G
PC302754-25G
PC302754-100G
APOSPC302754-5GEA
136
EUR
APOSPC302754-5G
APOSPC302754-1G
APOSPC302754-25G
APOSPC302754-100G
4-Fluoro-3,5-dimethylbenzoic acid 98%
4-Fluoro-3,5-dimethylbenzoic acid
Formel:
C₉H₉FO₂ Schmelzpunkt: 170…173 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00184969 CAS-Nummer: 120512-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...