4-Fluor-2-methylbenzylalkohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC302752-10G
PC302752-5G
APOSPC302752-10GEA
296
EUR
APOSPC302752-10G
APOSPC302752-5G
4-Fluor-2-methylbenzylalkohol 97%
4-Fluor-2-methylbenzylalkohol
Formel:
C₈H₉FO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03701058 CAS-Nummer: 80141-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...