5-Fluoro-3-methylindole
Lieferant: Apollo Scientific
|
Achtung
|
PC3027-1G
PC3027-250MG
APOSPC3027-1GEA
219
EUR
APOSPC3027-1G
APOSPC3027-250MG
5-Fluoro-3-methylindole
5-Fluoro-3-methylindole
Formel:
C₉H₈FN Schmelzpunkt: 85 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03095377 CAS-Nummer: 392-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...