2-Methyl-5-trifluoromethoxy-DL-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-5-trifluoromethoxy-DL-phenylalanine
PC302473-1G
PC302473-5G
APOSPC302473-1GEA
159
EUR
APOSPC302473-1G
APOSPC302473-5G
2-Methyl-5-trifluoromethoxy-DL-phenylalanine
2-Methyl-5-trifluoromethoxy-DL-phenylalanine
Formel:
C₁₁H₁₂F₃NO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22201059 CAS-Nummer: 1435806-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...