(2-Fluor-5-methylphenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Fluoro-5-methylphenyl)acetic acid

PC302388-1G PC302388-5G PC302388-250MG
APOSPC302388-1GEA 128 EUR
APOSPC302388-1G APOSPC302388-5G APOSPC302388-250MG
(2-Fluor-5-methylphenyl)essigsäure 98%
(2-Fluor-5-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Schmelzpunkt: 90…93 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06660230
CAS-Nummer: 203314-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR