4-Chloro-2-fluoro-3-methoxybenzyl alcohol 97%

Lieferant: Apollo Scientific

PC302324-1G PC302324-5G
APOSPC302324-1GEA 94.4 EUR
APOSPC302324-1G APOSPC302324-5G
4-Chloro-2-fluoro-3-methoxybenzyl alcohol 97%
4-Chloro-2-fluoro-3-methoxybenzyl alcohol
Formel: C₈H₈ClFO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19687181
CAS-Nummer: 1323966-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR