3-Chlor-5-(trifluormethoxy)anilin 98%

Lieferant: Apollo Scientific

PC302289-1G PC302289-5G PC302289-250MG
APOSPC302289-1GEA 94.4 EUR
APOSPC302289-1G APOSPC302289-5G APOSPC302289-250MG
3-Chlor-5-(trifluormethoxy)anilin 98%
3-Chlor-5-(trifluormethoxy)anilin
Formel: C₇H₅ClF₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09832245
CAS-Nummer: 151276-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR