3-(4-Fluor-3-methylphenyl)propionsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302266-1G
PC302266-5G
PC302266-10G
PC302266-25G
APOSPC302266-1GEA
72.5
EUR
APOSPC302266-1G
APOSPC302266-5G
APOSPC302266-10G
APOSPC302266-25G
3-(4-Fluor-3-methylphenyl)propionsäure 98%
3-(4-Fluor-3-methylphenyl)propionsäure
Formel:
C₁₀H₁₁FO₂ Schmelzpunkt: 64…69 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09025393 CAS-Nummer: 881189-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...