2-Methoxy-4-(trifluormethyl)phenylacetonitril 97%
Lieferant: Apollo Scientific
PC302252-1G
PC302252-5G
PC302252-10G
APOSPC302252-1GEA
159
EUR
APOSPC302252-1G
APOSPC302252-5G
APOSPC302252-10G
2-Methoxy-4-(trifluormethyl)phenylacetonitril 97%
2-Methoxy-4-(trifluormethyl)phenylacetonitril
Formel:
C₁₀H₈F₃NO Schmelzpunkt: 75…78 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832352 CAS-Nummer: 1017779-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...