2-Ethoxy-3,5-difluoranilin 98%
Lieferant: Apollo Scientific
PC302235-1G
PC302235-5G
PC302235-10G
APOSPC302235-1GEA
116
EUR
APOSPC302235-1G
APOSPC302235-5G
APOSPC302235-10G
2-Ethoxy-3,5-difluoranilin 98%
2-Ethoxy-3,5-difluoranilin
Formel:
C₈H₉F₂NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09258714 CAS-Nummer: 1017779-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...