4-Chloro-2-(trifluoromethoxy)benzamide 98%
Lieferant: Apollo Scientific
Synonyme:
4-chloro-2-(trifluoromethoxy)benzamide
PC302102-1G
PC302102-5G
APOSPC302102-1GEA
69.6
EUR
APOSPC302102-1G
APOSPC302102-5G
4-Chloro-2-(trifluoromethoxy)benzamide 98%
4-Chloro-2-(trifluoromethoxy)benzamide
Formel:
C₈H₃Cl₂F₃O₂ Molekulargewicht: 259.01 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18393778 CAS-Nummer: 1261791-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...