2-Methoxy-6-(trifluormethyl)benzylalkohol 98%
Lieferant: Apollo Scientific
PC302079-1G
PC302079-5G
PC302079-10G
APOSPC302079-1GEA
126
EUR
APOSPC302079-1G
APOSPC302079-5G
APOSPC302079-10G
2-Methoxy-6-(trifluormethyl)benzylalkohol 98%
2-Methoxy-6-(trifluormethyl)benzylalkohol
Formel:
C₉H₉F₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832325 CAS-Nummer: 1017779-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...