4-Ethoxy-2,6-difluorbenzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

PC302062-1G PC302062-5G
APOSPC302062-1GEA 79.9 EUR
APOSPC302062-1G APOSPC302062-5G
4-Ethoxy-2,6-difluorbenzylalkohol 97%
4-Ethoxy-2,6-difluorbenzylalkohol
Formel: C₉H₁₀F₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09258696
CAS-Nummer: 438050-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR