Ethyl 2-amino-4-trifluoromethylbenzoate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-amino-4-trifluoromethylbenzoate

PC300714-1G PC300714-5G
APOSPC300714-1GEA 126 EUR
APOSPC300714-1G APOSPC300714-5G
Ethyl 2-amino-4-trifluoromethylbenzoate 95%
Ethyl-2-amino-4-trifluoromethylbenzoate
Formel: C₁₀H₁₀F₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08274771
CAS-Nummer: 65568-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR