2-(1-Methylhydrazino)-4-(trifluoromethyl)pyrimidine
Lieferant: Apollo Scientific
Synonyme:
2-(1-Methylhydrazino)-4-(trifluoromethyl)pyrimidine
PC300693-1G
PC300693-10G
PC300693-5G
APOSPC300693-1GEA
217
EUR
APOSPC300693-1G
APOSPC300693-10G
APOSPC300693-5G
2-(1-Methylhydrazino)-4-(trifluoromethyl)pyrimidine
2-(1-Methylhydrazino)-4-(trifluoromethyl)pyrimidine
Formel:
C₆H₇F₃N₄ Molekulargewicht: 192.14 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20037418 CAS-Nummer: 1372711-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...