4-(4-Fluorophenoxy)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(4-Fluorophenoxy)phenyl)methanol

PC300610-1G PC300610-5G
APOSPC300610-1GEA 186 EUR
APOSPC300610-1G APOSPC300610-5G
4-(4-Fluorophenoxy)benzyl alcohol
4-(4-Fluorophenoxy)benzyl alcohol
Formel: C₁₃H₁₁FO₂
Schmelzpunkt: 66…68 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11190422
CAS-Nummer: 167091-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR