2,6-Diamino-3,5-difluorpyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC2831-1G
PC2831-5G
PC2831-25G
APOSPC2831-1GEA
0
EUR
APOSPC2831-1G
APOSPC2831-5G
APOSPC2831-25G
2,6-Diamino-3,5-difluorpyridin
2,6-Diamino-3,5-difluoropyridine
Formel:
C₅H₅F₂N₃ Schmelzpunkt: 150…160 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03412208 CAS-Nummer: 247069-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...