Pentafluorphenyl-1-ethylensulfonat 95%

Lieferant: Apollo Scientific

PC28081-1G PC28081-5G
APOSPC28081-1GEA 336 EUR
APOSPC28081-1G APOSPC28081-5G
Pentafluorphenyl-1-ethylensulfonat 95%
Pentafluorphenyl-1-ethylensulfonat
Formel: C₈H₃F₅O₃S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06659554
CAS-Nummer: 452905-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR