3-fluoro-5-iodopyridine

Lieferant: Apollo Scientific
Achtung

PC2676-1G PC2676-250MG
APOSPC2676-1GEA 29 EUR
APOSPC2676-1G APOSPC2676-250MG
3-fluoro-5-iodopyridine
3-Fluoro-5-iodopyridine
Formel: C₅H₃FIN
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08457780
CAS-Nummer: 757950-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR