3-Brom-4-(trifluormethoxy)phenol
Lieferant: Apollo Scientific
|
Achtung
|
PC2672-5G
PC2672-25G
APOSPC2672-5GEA
86.1
EUR
APOSPC2672-5G
APOSPC2672-25G
3-Brom-4-(trifluormethoxy)phenol
3-Brom-4-(trifluormethoxy)phenol
Formel:
C₇H₄BrF₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05864310 CAS-Nummer: 886496-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...