(2-Chlor-5-fluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-fluorophenylacetic acid

PC2515-10G PC2515-5G PC2515-1G
APOSPC2515-10GEA 139 EUR
APOSPC2515-10G APOSPC2515-5G APOSPC2515-1G
(2-Chlor-5-fluorphenyl)essigsäure 98%
(2-Chlor-5-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01016155
CAS-Nummer: 177985-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR