1-Brom-2-fluor-5-iodbenzol 98%

Lieferant: Apollo Scientific
Achtung

PC2414-100G PC2414-25G PC2414-5G
APOSPC2414-100GEA 109 EUR
APOSPC2414-100G APOSPC2414-25G APOSPC2414-5G
1-Brom-2-fluor-5-iodbenzol 98%
1-Brom-2-fluor-5-iodbenzol
Formel: C₆H₃BrFI
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD04116322
CAS-Nummer: 811842-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR