1,3-Dibrom-5-fluor-2-iodbenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dibromo-4-fluoroiodobenzene
PC2263M-5G
PC2263M-100G
PC2263M-25G
APOSPC2263M-5GEA
21.9
EUR
APOSPC2263M-5G
APOSPC2263M-100G
APOSPC2263M-25G
1,3-Dibrom-5-fluor-2-iodbenzol 98%
1,3-Dibrom-5-fluor-2-iodbenzol
Formel:
C₆H₂Br₂FI Schmelzpunkt: 128…130 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00042232 CAS-Nummer: 62720-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...