Fmoc-trans-4-fluoro-Pro-Oh

Lieferant: Apollo Scientific

Synonyme: (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid, N-FMOC protected, Fmoc-trans-4-F-Pro-OH

PC2201-100MG PC2201-1G PC2201-250MG
APOSPC2201-100MGEA 30.6 EUR
APOSPC2201-100MG APOSPC2201-1G APOSPC2201-250MG
Fmoc-trans-4-fluoro-Pro-Oh
Fmoc-trans-4-fluoro-Pro-Oh
Formel: C₂₀H₁₈FNO₄
Molekulargewicht: 355.365 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03424224
CAS-Nummer: 203866-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR