3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2-hydrazinyl-5-(trifluoromethyl)pyridine
PC2075JK-1G
PC2075JK-5G
PC2075JK-10G
APOSPC2075JK-1GEA
21.9
EUR
APOSPC2075JK-1G
APOSPC2075JK-5G
APOSPC2075JK-10G
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine
Formel:
C₆H₅ClF₃N₃ Schmelzpunkt: 90…94 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00067865 CAS-Nummer: 89570-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...