5-Fluorobenzo[d]isoxazol-3-amine 95+%

Lieferant: Apollo Scientific

PC201332-1G PC201332-5G
APOSPC201332-1GEA 171 EUR
APOSPC201332-1G APOSPC201332-5G
5-Fluorobenzo[d]isoxazol-3-amine 95+%
5-Fluorobenzo[d]isoxazol-3-amine
Formel: C7H5FN2O
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09999137
CAS-Nummer: 868271-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR