Ethyl-2-fluor-5-iodbenzoat 95+%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-Fluoro-5-iodobenzoate
PC201304-1G
PC201304-5G
APOSPC201304-1GEA
87
EUR
APOSPC201304-1G
APOSPC201304-5G
Ethyl-2-fluor-5-iodbenzoat 95+%
Ethyl-2-fluor-5-iodbenzoat
Formel:
C₉H₈FIO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06204769 CAS-Nummer: 773136-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...