5-Fluoro-2-pyrimidinecarbonitrile 97%
Lieferant: Apollo Scientific
Synonyme:
5-Fluoropyrimidine-2-carbonitrile
PC201244-10G
PC201244-1G
PC201244-5G
APOSPC201244-10GEA
808
EUR
APOSPC201244-10G
APOSPC201244-1G
APOSPC201244-5G
5-Fluoro-2-pyrimidinecarbonitrile 97%
5-Fluoro-2-pyrimidinecarbonitrile
Formel:
C5H2FN3 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11846243 CAS-Nummer: 38275-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...