2,4-Difluor-3-hydroxybenzaldehyd 95+%
Lieferant: Apollo Scientific
PC201189-1G
PC201189-5G
APOSPC201189-1GEA
135
EUR
APOSPC201189-1G
APOSPC201189-5G
2,4-Difluor-3-hydroxybenzaldehyd 95+%
2,4-Difluoro-3-hydroxybenzaldehyde
Formel:
C₇H₄F₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12405741 CAS-Nummer: 192927-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...