2,3-Dichloro-4-fluorobenzoic acid 98%

Lieferant: Apollo Scientific

PC201150-10G PC201150-1G PC201150-5G
APOSPC201150-10GEA 516 EUR
APOSPC201150-10G APOSPC201150-1G APOSPC201150-5G
2,3-Dichloro-4-fluorobenzoic acid 98%
2,3-Dichloro-4-fluorobenzoic acid
Formel: C₇H₃Cl₂FO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09835200
CAS-Nummer: 154257-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR