Methyl-4-fluor-2-methoxybenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-2-methoxybenzoic acid methyl ester, Methyl 4-fluoro-2-methoxybenzoate
PC200606-1G
PC200606-5G
PC200606-10G
APOSPC200606-1GEA
21.9
EUR
APOSPC200606-1G
APOSPC200606-5G
APOSPC200606-10G
Methyl-4-fluor-2-methoxybenzoat
Methyl-4-fluor-2-methoxybenzoat
Formel:
C₉H₉FO₃ Siedepunkt: 78…80 °C/0.05mmHg Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06203990 CAS-Nummer: 204707-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...