Methyl-2-amino-4-fluoro-5-methoxybenzoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-4-fluoro-5-methoxybenzoate
PC200348-1G
PC200348-500MG
APOSPC200348-1GEA
361
EUR
APOSPC200348-1G
APOSPC200348-500MG
Methyl-2-amino-4-fluoro-5-methoxybenzoate
Methyl-2-amino-4-fluoro-5-methoxybenzoate
Formel:
C₉H₁₀FNO₃ Schmelzpunkt: 43…46 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16159159 CAS-Nummer: 159768-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...