Methyl-2-bromo-4-fluoro-5-methoxybenzoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-bromo-4-fluoro-5-methoxybenzoate
PC200333-1G
PC200333-500MG
APOSPC200333-1GEA
181
EUR
APOSPC200333-1G
APOSPC200333-500MG
Methyl-2-bromo-4-fluoro-5-methoxybenzoate
Methyl-2-bromo-4-fluoro-5-methoxybenzoate
Formel:
C₉H₈BrFO₃ Schmelzpunkt: 36…38 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20482372 CAS-Nummer: 1007455-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...