N-Methyl-2-(trifluoromethyl)-4-quinazolinamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Methylamino)-2-(trifluoromethyl)quinazoline
PC200293-1G
PC200293-500MG
APOSPC200293-1GEA
626
EUR
APOSPC200293-1G
APOSPC200293-500MG
N-Methyl-2-(trifluoromethyl)-4-quinazolinamine
N-Methyl-2-(trifluoromethyl)-4-quinazolinamine
Formel:
C₁₀H₈F₃N₃ Schmelzpunkt: 143…144 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08762128 CAS-Nummer: 929418-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...