Ethyl 2-phenoxy-5-(trifluoromethyl)nicotinate

Lieferant: Apollo Scientific

PC200286-1G PC200286-500MG
APOSPC200286-1GEA 471 EUR
APOSPC200286-1G APOSPC200286-500MG
Ethyl 2-phenoxy-5-(trifluoromethyl)nicotinate
Ethyl-2-phenoxy-5-(trifluoromethyl)nicotinate
Formel: C₁₅H₁₂F₃NO₃
Schmelzpunkt: 48…50 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13193104
CAS-Nummer: 1192263-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR