Potassium-2-thienyltrifluoroborate

Lieferant: Apollo Scientific

Synonyme: Potassium 2-Thienyltrifluoroborate

PC200178-1G PC200178-500MG PC200178-5G
APOSPC200178-1GEA 102 EUR
APOSPC200178-1G APOSPC200178-500MG APOSPC200178-5G
Potassium-2-thienyltrifluoroborate
Potassium-2-thienyltrifluoroborate
Formel: C₄H₃BF₃KS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04112719
CAS-Nummer: 906674-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR