2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol

Lieferant: Apollo Scientific

Synonyme: {2-[(4-Fluorophenyl)dimethylsilyl]phenyl}methanol

PC200130-1G PC200130-500MG PC200130-5G
APOSPC200130-1GEA 118 EUR
APOSPC200130-1G APOSPC200130-500MG APOSPC200130-5G
2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
Formel: C₁₅H₁₇FOSi
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16251550
CAS-Nummer: 853955-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR